Assay Details
| Gene Symbol: CACUL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003483565.4 | XP_003483613.3 | 690 | 92 | ||
| GenBank mRNA | AK393622.1 | - | 1 - 2 | 624 | 92 | |
More Information
Set Membership: |
|

CACUL1
Genomic Map