Assay Details
| Gene Symbol: MTREX | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003133964.6 | XP_003134012.3 | 3354 | 76 | ||
| GenBank mRNA | AK234447.1 | - | 26 - 27 | 3266 | 76 | |
More Information
Set Membership: |
|

MTREX
Genomic Map