Assay Details
| Gene Symbol: DCUN1D5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003129809.4 | XP_003129857.1 | 576 | 92 | ||
| XM_021062713.1 | XP_020918372.1 | 578 | 92 | |||
| XM_021062714.1 | XP_020918373.1 | 489 | 92 | |||
| XM_021062715.1 | XP_020918374.1 | 484 | 92 | |||
| GenBank mRNA | AK239201.1 | - | 3 - 4 | 567 | 92 | |
| AY610026.1 | - | 3 - 4 | 466 | 92 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021062714.1, XM_021062715.1 transcripts.
Set Membership: |
|

DCUN1D5
Genomic Map