Assay Details
| Gene Symbol: CBX6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021091475.1 | XP_020947134.1 | 214 | 68 | ||
| XM_021091476.1 | XP_020947135.1 | 166 | 68 | |||
| GenBank mRNA | AK390394.1 | - | 1 - 2 | 155 | 68 | |
| AK400169.1 | - | 1 - 2 | 154 | 68 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021091476.1 transcripts.
Set Membership: |
|

CBX6
Genomic Map