Assay Details
Gene Symbol: REXO2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003129878.3 | XP_003129926.1 | 299 | 78 | ||
GenBank mRNA | AK347351.1 | - | 1 - 2 | 297 | 78 | |
AK394948.1 | - | 1 - 2 | 199 | 78 | ||
AY609650.1 | - | 1 - 2 | 171 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
