Assay Details
| Gene Symbol: ENSA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_214174.2 | NP_999339.2 | 3 - 4 | 486 | 52 | |
| GenBank mRNA | AK239443.1 | - | 3 - 4 | 626 | 52 | |
| AK396895.1 | - | 3 - 4 | 485 | 52 | ||
| AY609423.1 | - | 3 - 4 | 468 | 52 | ||
| BP142411.1 | - | 3 - 4 | 557 | 52 | ||
| CN162636.1 | - | 1 - 2 | 585 | 52 | ||
More Information
Set Membership: |
|

ENSA
Genomic Map