Assay Details
| Gene Symbol: RPS3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044601.1 | NP_001038066.1 | 4 - 5 | 351 | 80 | |
| XM_021102267.1 | XP_020957926.1 | 869 | 80 | |||
| XM_021102268.1 | XP_020957927.1 | 777 | 80 | |||
| XM_021102269.1 | XP_020957928.1 | 777 | 80 | |||
| GenBank mRNA | AK234402.1 | - | 6 - 7 | 850 | 80 | |
| AK390990.1 | - | 6 - 7 | 783 | 80 | ||
| AK392066.1 | - | 4 - 5 | 377 | 80 | ||
| AK398968.1 | - | 4 - 5 | 394 | 80 | ||
| DQ660373.1 | - | 4 - 5 | 351 | 80 | ||
More Information
Set Membership: |
|

RPS3
Genomic Map