Assay Details
| Gene Symbol: UBE2G1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021067700.1 | XP_020923359.1 | 916 | 105 | ||
| GenBank mRNA | AK233312.1 | - | 5 - 6 | 869 | 105 | |
| AK239081.1 | - | 5 - 6 | 886 | 105 | ||
| AK397752.1 | - | 5 - 6 | 915 | 105 | ||
| AY610214.1 | - | 5 - 6 | 871 | 105 | ||
More Information
Set Membership: |
|

UBE2G1
Genomic Map