Assay Details
Gene Symbol: AUTS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001246269.1 | NP_001233198.1 | 13 - 14 | 1951 | 67 | |
GenBank mRNA | AB570017.1 | - | 13 - 14 | 1951 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
