Assay Details
Gene Symbol: PLPP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001924179.5 | XP_001924214.1 | 418 | 79 | ||
GenBank mRNA | AK238542.1 | - | 4 - 5 | 422 | 79 | |
AK238550.1 | - | 4 - 5 | 424 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
