Assay Details
| Gene Symbol: BCL11A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001297634.1 | NP_001284563.1 | 1 - 2 | 106 | 72 | |
| XM_013996120.2 | XP_013851574.1 | 494 | 72 | |||
| XM_021085693.1 | XP_020941352.1 | 533 | 72 | |||
| XM_021085694.1 | XP_020941353.1 | 811 | 72 | |||
| XM_021085696.1 | XP_020941355.1 | 887 | 72 | |||
| XM_021085697.1 | XP_020941356.1 | 537 | 72 | |||
| XM_021085698.1 | XP_020941357.1 | 2292 | 72 | |||
| XM_021085699.1 | XP_020941358.1 | 536 | 72 | |||
| XM_021085702.1 | XP_020941361.1 | 811 | 72 | |||
| GenBank mRNA | AK231444.1 | - | 1 - 2 | 355 | 72 | |
| AY609448.1 | - | 1 - 2 | 269 | 72 | ||
| KJ652909.1 | - | 1 - 2 | 106 | 72 | ||
More Information
Set Membership: |
|

BCL11A
Genomic Map