Assay Details
Gene Symbol: SEC61B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243825.1 | NP_001230754.1 | 3 - 4 | 299 | 88 | |
GenBank mRNA | AK236961.1 | - | 3 - 4 | 299 | 88 | |
AK343873.1 | - | 3 - 4 | 314 | 88 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
