Assay Details
| Gene Symbol: DPY30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244925.1 | NP_001231854.1 | 4 - 5 | 316 | 124 | |
| XM_005655249.3 | XP_005655306.1 | 455 | 124 | |||
| GenBank mRNA | AK346618.1 | - | 4 - 5 | 316 | 124 | |
| AK392665.1 | - | 4 - 5 | 317 | 124 | ||
More Information
Set Membership: |
|

DPY30
Genomic Map