Assay Details
Gene Symbol: MYL6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163997.1 | NP_001157469.1 | 5 - 6 | 459 | 64 | |
GenBank mRNA | AK238248.1 | - | 5 - 6 | 458 | 64 | |
AK240360.1 | - | 5 - 6 | 459 | 64 | ||
AK391962.1 | - | 5 - 6 | 466 | 64 | ||
AK397509.1 | - | 5 - 6 | 460 | 64 | ||
AK399933.1 | - | 5 - 6 | 472 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
