Assay Details
Gene Symbol: WTAP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244241.1 | NP_001231170.1 | 5 - 6 | 459 | 65 | |
NM_001244242.1 | NP_001231171.1 | 5 - 6 | 443 | 65 | ||
XM_005659114.3 | XP_005659171.1 | 478 | 65 | |||
XM_021101793.1 | XP_020957452.1 | 478 | 65 | |||
XM_021101851.1 | XP_020957510.1 | 600 | 65 | |||
GenBank mRNA | AK231140.1 | - | 6 - 7 | 343 | 65 | |
AK233198.1 | - | 5 - 6 | 433 | 65 | ||
AK236344.1 | - | 5 - 6 | 443 | 65 | ||
AK237427.1 | - | 6 - 7 | 340 | 65 | ||
AK351349.1 | - | 5 - 6 | 459 | 65 | ||
AK391167.1 | - | 5 - 6 | 443 | 65 | ||
AK394413.1 | - | 5 - 6 | 430 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
