Assay Details
Gene Symbol: YTHDF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005665152.3 | XP_005665209.1 | 89 | |||
GenBank mRNA | AK235654.1 | - | 3 - 4 | 461 | 89 | |
AK238769.1 | - | 3 - 4 | 89 | |||
AK396412.1 | - | 3 - 4 | 89 | |||
AK398480.1 | - | 3 - 4 | 89 | |||
AK399273.1 | - | 1 - 2 | 89 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
