Assay Details
Gene Symbol: SMU1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243898.1 | NP_001230827.1 | 5 - 6 | 663 | 96 | |
GenBank mRNA | AK343711.1 | - | 5 - 6 | 663 | 96 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
