Assay Details
| Gene Symbol: SLC25A4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003133345.4 | XP_003133393.1 | 463 | 66 | ||
| GenBank mRNA | AK233419.1 | - | 1 - 2 | 244 | 66 | |
| AK392539.1 | - | 1 - 2 | 215 | 66 | ||
More Information
Set Membership: |
|

SLC25A4
Genomic Map