Assay Details
Gene Symbol: MEMO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244877.1 | NP_001231806.1 | 3 - 4 | 608 | 70 | |
XM_005655246.3 | XP_005655303.1 | 403 | 70 | |||
XM_005655247.3 | XP_005655304.1 | 354 | 70 | |||
XM_021085669.1 | XP_020941328.1 | 73 | 70 | |||
GenBank mRNA | AK343321.1 | - | 3 - 4 | 608 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
