Assay Details
Gene Symbol: ATP5F1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001185142.1 | NP_001172071.1 | 6 - 7 | 844 | 68 | |
GenBank mRNA | AK232528.1 | - | 6 - 7 | 849 | 68 | |
AK236714.1 | - | 6 - 7 | 861 | 68 | ||
AK238748.1 | - | 7 - 8 | 740 | 68 | ||
AK396284.1 | - | 6 - 7 | 861 | 68 | ||
AK396428.1 | - | 6 - 7 | 859 | 68 | ||
AK396740.1 | - | 6 - 7 | 849 | 68 | ||
AK396809.1 | - | 6 - 7 | 884 | 68 | ||
AK397056.1 | - | 6 - 7 | 849 | 68 | ||
AK398493.1 | - | 3 - 4 | 427 | 68 | ||
AY609645.1 | - | 6 - 7 | 845 | 68 | ||
EU650784.1 | - | 6 - 7 | 844 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
