Assay Details
| Gene Symbol: FITM2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001128460.1 | NP_001121932.1 | 1 - 2 | 182 | 70 | |
| GenBank mRNA | EU596460.1 | - | 1 - 2 | 182 | 70 | |
| FJ393219.1 | - | 1 - 2 | 254 | 70 | ||
| JN630475.1 | - | 1 - 2 | 173 | 70 | ||
| KR132252.1 | - | 1 - 2 | 173 | 70 | ||
More Information
Set Membership: |
|

FITM2
Genomic Map