Assay Details
| Gene Symbol: CDK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001159304.2 | NP_001152776.1 | 4 - 5 | 433 | 84 | |
| XM_005671013.3 | XP_005671070.1 | 443 | 84 | |||
| XM_005671016.3 | XP_005671073.1 | 363 | 84 | |||
| GenBank mRNA | AB045783.1 | - | 1 - 2 | 112 | 84 | |
| AB495208.1 | - | 3 - 4 | 343 | 84 | ||
| AK238864.1 | - | 4 - 5 | 432 | 84 | ||
| BP161545.1 | - | 4 - 5 | 433 | 84 | ||
| GQ184633.1 | - | 4 - 5 | 396 | 84 | ||
More Information
Set Membership: |
|

CDK1
Genomic Map