Assay Details
Gene Symbol: OSTM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001927476.5 | XP_001927511.1 | 548 | 67 | ||
GenBank mRNA | AB271955.1 | - | 1 - 2 | 400 | 67 | |
AK396352.1 | - | 1 - 2 | 487 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
