Assay Details
Gene Symbol: CALM3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244209.1 | NP_001231138.1 | 4 | 426 | 77 | |
GenBank mRNA | AF034988.1 | - | 2 | 150 | 77 | |
AK234945.1 | - | 4 | 417 | 77 | ||
AK391956.1 | - | 4 | 427 | 77 | ||
AK392323.1 | - | 4 | 364 | 77 | ||
AK400416.1 | - | 4 | 408 | 77 | ||
FS666815.1 | - | 4 | 427 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
