Assay Details
| Gene Symbol: COL8A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001926443.6 | XP_001926478.1 | 2571 | 60 | ||
| XM_005670255.3 | XP_005670312.1 | 2484 | 60 | |||
| GenBank mRNA | AF054891.1 | - | 1 | 454 | 60 | |
More Information
Set Membership: |
|

COL8A1
Genomic Map