Assay Details
Gene Symbol: HOXA10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244338.2 | NP_001231267.1 | 2 | 375 | 105 | |
NM_001257354.2 | NP_001244283.1 | 2 | 1130 | 105 | ||
NR_155016.1 | 3 | 549 | 105 | |||
NR_155017.1 | 3 | 611 | 105 | |||
XM_005673290.3 | XP_005673347.1 | 2585 | 105 | |||
XM_005673291.3 | XP_005673348.1 | 2647 | 105 | |||
XM_005673294.3 | XP_005673351.1 | 957 | 105 | |||
XM_005673295.3 | XP_005673352.1 | 669 | 105 | |||
XM_013985769.2 | XP_013841223.1 | 1499 | 105 | |||
XM_013985770.2 | XP_013841224.1 | 1437 | 105 | |||
XM_021078489.1 | XP_020934148.1 | 5186 | 105 | |||
XM_021078491.1 | XP_020934150.1 | 1123 | 105 | |||
XM_021078493.1 | XP_020934152.1 | 3558 | 105 | |||
GenBank mRNA | AK396478.1 | - | 2 | 1072 | 105 | |
HQ700342.1 | - | 2 | 315 | 105 | ||
JN836600.1 | - | 2 | 1130 | 105 | ||
KT823698.1 | - | 3 | 551 | 105 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
