Assay Details
| Gene Symbol: GFAP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244397.1 | NP_001231326.1 | 4 - 5 | 784 | 54 | |
| XM_005668709.3 | XP_005668766.1 | 797 | 54 | |||
| GenBank mRNA | AJ551395.1 | - | 3 - 4 | 314 | 54 | |
| AK392124.1 | - | 4 - 5 | 785 | 54 | ||
| BP997870.1 | - | 2 - 3 | 191 | 54 | ||
More Information
Set Membership: |
|

GFAP
Genomic Map