Assay Details
Gene Symbol: CNN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003125902.5 | XP_003125950.3 | 712 | 60 | ||
GenBank mRNA | AK234188.1 | - | 5 - 6 | 728 | 60 | |
AK396406.1 | - | 5 - 6 | 728 | 60 | ||
AM490167.1 | - | 1 - 2 | 67 | 60 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
