Assay Details
Gene Symbol: BGLAP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164004.1 | NP_001157476.1 | 4 | 356 | 120 | |
XM_013996978.2 | XP_013852432.1 | 561 | 120 | |||
GenBank mRNA | AY150038.1 | - | 1 | 147 | 120 | |
DV224135.1 | - | 4 | 374 | 120 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
