Assay Details
| Gene Symbol: RPL12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005660442.3 | XP_005660499.1 | 157 | 73 | ||
| GenBank mRNA | AK237253.1 | - | 1 - 2 | 127 | 73 | |
| AK345030.1 | - | 1 - 2 | 135 | 73 | ||
| AK394515.1 | - | 1 - 2 | 131 | 73 | ||
| AK397580.1 | - | 1 - 2 | 127 | 73 | ||
| AY550045.1 | - | 1 - 2 | 117 | 73 | ||
| AY610349.1 | - | 1 - 2 | 132 | 73 | ||
More Information
Set Membership: |
|

RPL12
Genomic Map