Assay Details
Gene Symbol: ADORA2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003483462.4 | XP_003483510.2 | 2298 | 67 | ||
GenBank mRNA | AY686733.1 | - | 1 - 2 | 276 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
