Assay Details
Gene Symbol: P2RY1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005669935.3 | XP_005669992.1 | 1563 | 144 | ||
XM_005669936.3 | XP_005669993.1 | 1201 | 144 | |||
GenBank mRNA | AK231207.1 | - | 1 | 1258 | 144 | |
AK233663.1 | - | 1 | 1532 | 144 | ||
AK348897.1 | - | 1 | 1325 | 144 | ||
AY609398.1 | - | 1 | 721 | 144 | ||
AY691680.1 | - | 1 | 655 | 144 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
