Assay Details
| Gene Symbol: LPAR4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013986253.2 | XP_013841707.1 | 1497 | 155 | ||
| XM_013986254.2 | XP_013841708.1 | 1610 | 155 | |||
| XM_021080213.1 | XP_020935872.1 | 1497 | 155 | |||
| XM_021080214.1 | XP_020935873.1 | 1547 | 155 | |||
| GenBank mRNA | EF183526.1 | - | 1 | 354 | 155 | |
More Information
Set Membership: |
|

LPAR4
Genomic Map