Assay Details
| Gene Symbol: RDH10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021089230.1 | XP_020944889.1 | 507 | 93 | ||
| GenBank mRNA | EU056577.1 | - | 1 - 2 | 205 | 93 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: RDH10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021089230.1 | XP_020944889.1 | 507 | 93 | ||
| GenBank mRNA | EU056577.1 | - | 1 - 2 | 205 | 93 | |
Set Membership: |
|