Assay Details
Gene Symbol: AQP10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001128454.1 | NP_001121926.1 | 4 - 5 | 491 | 69 | |
XM_013997036.2 | XP_013852490.1 | 827 | 69 | |||
GenBank mRNA | EU582021.1 | - | 4 - 5 | 491 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
