Assay Details
| Gene Symbol: RARA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003131473.4 | XP_003131521.1 | 1014 | 82 | ||
| XM_003131474.6 | XP_003131522.1 | 1117 | 82 | |||
| XM_021067021.1 | XP_020922680.1 | 951 | 82 | |||
| GenBank mRNA | AK395242.1 | - | 4 | 420 | 82 | |
| U82260.1 | - | 2 | 149 | 82 | ||
More Information
Set Membership: |
|

RARA
Genomic Map