Assay Details
| Gene Symbol: GJA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244212.1 | NP_001231141.1 | 2 | 1070 | 64 | |
| GenBank mRNA | AJ293888.1 | - | 1 | 328 | 64 | |
| EW652303.2 | - | 1 | 65 | 64 | ||
| X86023.1 | - | 151 | 64 | |||
More Information
Set Membership: |
|

GJA1
Genomic Map