Assay Details
Gene Symbol: PPP2R2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164020.1 | NP_001157492.1 | 6 - 7 | 925 | 78 | |
XM_005670440.3 | XP_005670497.1 | 728 | 78 | |||
XM_013982663.2 | XP_013838117.1 | 885 | 78 | |||
XM_013982664.2 | XP_013838118.1 | 744 | 78 | |||
XM_021072095.1 | XP_020927754.1 | 944 | 78 | |||
XM_021072096.1 | XP_020927755.1 | 523 | 78 | |||
GenBank mRNA | AK235933.1 | - | 6 - 7 | 921 | 78 | |
AK392519.1 | - | 6 - 7 | 925 | 78 | ||
Z34932.1 | - | 565 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
