Assay Details
Gene Symbol: GEM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001926976.7 | XP_001927011.3 | 761 | 67 | ||
XM_005662965.3 | XP_005663022.1 | 1706 | 67 | |||
GenBank mRNA | Z80109.1 | - | 685 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
