Assay Details
| Gene Symbol: LOC396684 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AJ293892.1 | - | 3 | 455 | 97 | |
| AK232297.1 | - | 5 | 1104 | 97 | ||
| M29508.1 | - | 5 | 1064 | 97 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC396684| Gene Symbol: LOC396684 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AJ293892.1 | - | 3 | 455 | 97 | |
| AK232297.1 | - | 5 | 1104 | 97 | ||
| M29508.1 | - | 5 | 1064 | 97 | ||
Set Membership: |
|