Assay Details
| Gene Symbol: CXCR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021075280.1 | XP_020930939.1 | 1040 | 72 | ||
| XM_021075281.1 | XP_020930940.1 | 1027 | 72 | |||
| XM_021075282.1 | XP_020930941.1 | 3424 | 72 | |||
| XM_021075283.1 | XP_020930942.1 | 930 | 72 | |||
| XM_021075284.1 | XP_020930943.1 | 943 | 72 | |||
| XM_021075285.1 | XP_020930944.1 | 3321 | 72 | |||
| GenBank mRNA | AK230703.1 | - | 3 | 948 | 72 | |
More Information
Set Membership: |
|

CXCR2
Genomic Map