Assay Details
| Gene Symbol: RPS16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001137620.1 | NP_001131092.1 | 5 | 397 | 60 | |
| GenBank mRNA | AK240126.1 | - | 921 | 60 | ||
| AK350281.1 | - | 5 | 437 | 60 | ||
| AY550068.1 | - | 2 | 148 | 60 | ||
| AY610244.1 | - | 5 | 452 | 60 | ||
| FJ268644.1 | - | 5 | 397 | 60 | ||
More Information
Set Membership: |
|

RPS16
Genomic Map