Assay Details
| Gene Symbol: DEGS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244121.1 | NP_001231050.1 | 2 | 362 | 67 | |
| XM_005653768.3 | XP_005653825.1 | 387 | 67 | |||
| GenBank mRNA | AK236774.1 | - | 2 | 364 | 67 | |
| AY609708.1 | - | 2 | 343 | 67 | ||
More Information
Set Membership: |
|

DEGS1
Genomic Map