Assay Details
| Gene Symbol: CX3CL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021093908.1 | XP_020949567.1 | 1170 | 57 | ||
| GenBank mRNA | DQ991100.1 | - | 1 | 53 | 57 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: CX3CL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021093908.1 | XP_020949567.1 | 1170 | 57 | ||
| GenBank mRNA | DQ991100.1 | - | 1 | 53 | 57 | |
Set Membership: |
|