Assay Details
| Gene Symbol: CERS6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021075469.1 | XP_020931128.1 | 847 | 94 | ||
| XM_021075470.1 | XP_020931129.1 | 893 | 94 | |||
| XM_021075471.1 | XP_020931130.1 | 845 | 94 | |||
| GenBank mRNA | AK349159.1 | - | 7 | 846 | 94 | |
| EU153261.1 | - | 7 | 553 | 94 | ||
More Information
Set Membership: |
|

CERS6
Genomic Map