Assay Details
| Gene Symbol: HTR4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001001267.1 | NP_001001267.1 | 7 | 1143 | 84 | |
| GenBank mRNA | AY496868.2 | - | 7 | 1143 | 84 | |
| AY566638.1 | - | 7 | 1143 | 84 | ||
More Information
Set Membership: |
|

HTR4
Genomic Map