Assay Details
| Gene Symbol: GLRB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001001545.1 | NP_001001545.1 | 9 | 1350 | 113 | |
| XM_021100485.1 | XP_020956144.1 | 1552 | 113 | |||
| XM_021100486.1 | XP_020956145.1 | 1576 | 113 | |||
| XM_021100487.1 | XP_020956146.1 | 1556 | 113 | |||
| XM_021100488.1 | XP_020956147.1 | 1476 | 113 | |||
| XM_021100490.1 | XP_020956149.1 | 1536 | 113 | |||
| GenBank mRNA | AJ715855.1 | - | 9 | 1350 | 113 | |
More Information
Set Membership: |
|

GLRB
Genomic Map