Assay Details
| Gene Symbol: CCR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001001621.1 | NP_001001621.1 | 2 | 1155 | 120 | |
| GenBank mRNA | AB119268.1 | - | 2 | 1155 | 120 | |
| AK391265.1 | - | 2 | 1097 | 120 | ||
More Information
Set Membership: |
|

CCR1
Genomic Map