Assay Details
| Gene Symbol: MAOB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001001864.1 | NP_001001864.1 | 15 | 1549 | 74 | |
| XM_013986071.2 | XP_013841525.2 | 1599 | 74 | |||
| GenBank mRNA | AK232951.1 | - | 15 | 1533 | 74 | |
| AY596820.1 | - | 15 | 1549 | 74 | ||
More Information
Set Membership: |
|

MAOB
Genomic Map