Assay Details
| Gene Symbol: RNF114 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001001869.1 | NP_001001869.1 | 5 - 6 | 650 | 74 | |
| GenBank mRNA | AK233272.1 | - | 5 - 6 | 643 | 74 | |
| AY606064.1 | - | 5 - 6 | 650 | 74 | ||
More Information
Set Membership: |
|

RNF114
Genomic Map